Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Reaction Buffer for phi29 DNA Polymerase (10X)

Catalog No. B62
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B62 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B62 Supplier Thermo Scientific™ Supplier No. B62
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ 10X Buffer for phi29 DNA Polymerase is the optimal buffer recommended for use with highly processive phi29 DNA polymerase.

Thermo Scientific™ 10X Buffer for phi29 DNA Polymerase is the optimal buffer recommended for use with highly processive phi29 DNA polymerase.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Reaction Buffer
For Use With (Application) Isothermal Amplification
Quantity 5 x 1 mL
Research Category Traditional Cloning
Content And Storage

• 10X Reaction buffer for Phi29 DNA Polymerase (5 x 1.0 mL)

Store at –5 to –30°C.

Shipping Condition Dry Ice
Concentration 10X

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.