Learn More
Sigma Aldrich Reaction Chamber Chip - 20 ?l Fluidic 556 Cop
Catalog No. 9215481EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
92-154-81EA 1 Ea.
1 options
Catalog No. 92-154-81EA Supplier Sigma Aldrich Supplier No. 9215481EA
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Microfluidic generation of droplets can produce highly monodispersed droplets with high frequency (up to hundreds of kHz). Interest in droplet-based microfluidic systems has grown substantially, because microfluidics offers the ability to handle very small volumes (mul to fl) of fluids, provides better mixing, encapsulation, sorting, and sensing. Microfluidics can be used for high throughput experimentation. Microfluidic-based droplets have many diverse and varied applications such as particle synthesis and chemical analysis. Highly controlled droplet production also makes single cell analysis, or drug testing possible.Reaction chamber chip - 20 mul, Fluidic 556, COP is made of COP (Cyclic olefin polymer ). This reaction chamber chip is veratile and simple to use, and can be used for basic reactions to cell culture.
Specifications
Quantity 1 Ea.
Synonym Microfluidic chip