Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Reaction Coil, 1000 μL, Polytetrafluoroethylene, 120 °C, 500 psi

Catalog No. NC2827582 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This reaction coil is for liquid chromatography applications requiring on-line chemical modification or derivatization of analytes. It features an inert fluid path compatible with most solvents and reagents, even at elevated temperatures, and a knitted open tube design to maintain analytical resolution. It is used in Post Column Reaction Modules.

  • Volume: 1000 μL.
  • Coil material: Polytetrafluoroethylene (0.018 in. ID).
  • Fittings material: PEEK; Jacket material: Anodized aluminum.
  • Max temperature: 120 °C; Max pressure: 500 psi (3.4 MPa).
  • Inert fluid path for broad solvent compatibility and minimal band-spreading.
  • Maintains analytical resolution.
  • Includes 316 stainless steel junction tee.
  • Tee fitting provides temperature control and coil protection for Alliance HPLC Systems.

Catalog No. NC2827582 Supplier Waters Corp Supplier No. WAT030844
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This reaction coil is for liquid chromatography applications requiring on-line chemical modification or derivatization of analytes. It features an inert fluid path compatible with most solvents and reagents, even at elevated temperatures, and a knitted open tube design to maintain analytical resolution. It is used in Post Column Reaction Modules.

  • Volume: 1000 μL.
  • Coil material: Polytetrafluoroethylene (0.018 in. ID).
  • Fittings material: PEEK; Jacket material: Anodized aluminum.
  • Max temperature: 120 °C; Max pressure: 500 psi (3.4 MPa).
  • Inert fluid path for broad solvent compatibility and minimal band-spreading.
  • Maintains analytical resolution.
  • Includes 316 stainless steel junction tee.
  • Tee fitting provides temperature control and coil protection for Alliance HPLC Systems.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.