Learn More
DWK Life Sciences KIMBLE™ Reaction Flask Clamp, 4 in.
SDP
Catalog No. 12141418 Shop All DWK Life Sciences Products
Catalog No. 12-141-418 Supplier DWK Life Sciences Supplier No. 6137500000
Add to Cart
Add to Cart
KIMBLE™ Reaction Flask Clamp, 4 in. features cast aluminum construction to connect tops to flasks. Wing nuts or thumb screws are evenly spaced to assure uniform compression for a reliable, secure seal.
  • Cast aluminum body construction connects tops to flasks securely
  • Wing nuts or thumb screws are spaced to assure uniform compression for sealing the system
  • Fits ground flange ID of 4 inches
  • Compatible with 4-inch cylindrical and jacketed reaction flasks
TRUSTED_SUSTAINABILITY
Specifications
Product Type Reaction Flask Clamp
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Sterility Non-sterile