Learn More
Chemglass Life Sciences Reaction Heating Block, Anodized Clear, Fits (10) 5mm NMR Tubes
SDP
Catalog No. NC1424215
Encompass_Preferred
Reaction Heating Block, Anodized Clear, Fits (10) 5mm NMR Tubes. NMR Heating Reaction block is designed to rest on a round or square top hot plate stirrers. NMR heating block is an ideal replacement for oil baths. Block provides a safe and clean way for preheating NMR tubes, performing kinetics and degradation studies in 5mm NMR tubes. All blocks are anodized aluminum. A thermowell is provided for insertion of the temperature sensor.
Catalog No. NC1424215 Supplier Chemglass Life Sciences Supplier No. CG1991N10
Reaction Heating Block, Anodized Clear, Fits (10) 5mm NMR Tubes. NMR Heating Reaction block is designed to rest on a round or square top hot plate stirrers. NMR heating block is an ideal replacement for oil baths. Block provides a safe and clean way for preheating NMR tubes, performing kinetics and degradation studies in 5mm NMR tubes. All blocks are anodized aluminum. A thermowell is provided for insertion of the temperature sensor.