Learn More
Chemglass Life Sciences Reaction Vessel Lid, Stainless Steel, 300mm, 7-Neck, 106mm Center to Center Spacing on Necks, Viton O-Rings
SDP
Catalog No. CG1968B01
Encompass_Preferred
Reaction Vessel Lid, Stainless Steel, 300mm, 7-Neck, 106mm Center to Center Spacing on necks. Reaction vessel lid is constructed of 1/4 thick #316 Stainless Steel with a 1/8 thick PTFE gasket underneath. The only materials exposed to the reaction are PTFE and Viton. Lid has seven openings that accept 45/50 outer PTFE inserts. Inserts are secured to the lid using glass filled PTFE nuts. Lid can be used on any of our CG-1968 or CG-1972 reaction vessels. The flat design reduces the height 200mm than the standard glass reaction vessel lids.PLEASE NOTE: Stainless Steel HEAD DESIGNED FOR USE WITH CG-1968-X AND CG-1968-F SUPPORT FRAMES, AND NOT FOR USE WITH CG-1972-F SUPPORT FRAME.PLEASE NOTE: SPECIAL NECK SIZES AND CONFIGURATIONS WITH OPTIONAL O-RING MATERIALS ARE AVAILABLE. PLEASE CONTACT US FOR DETAILS.
Catalog No. CG1968B01 Supplier Chemglass Life Sciences Supplier No. CG1968B01
Reaction Vessel Lid, Stainless Steel, 300mm, 7-Neck, 106mm Center to Center Spacing on necks. Reaction vessel lid is constructed of 1/4 thick #316 Stainless Steel with a 1/8 thick PTFE gasket underneath. The only materials exposed to the reaction are PTFE and Viton. Lid has seven openings that accept 45/50 outer PTFE inserts. Inserts are secured to the lid using glass filled PTFE nuts. Lid can be used on any of our CG-1968 or CG-1972 reaction vessels. The flat design reduces the height 200mm than the standard glass reaction vessel lids.PLEASE NOTE: Stainless Steel HEAD DESIGNED FOR USE WITH CG-1968-X AND CG-1968-F SUPPORT FRAMES, AND NOT FOR USE WITH CG-1972-F SUPPORT FRAME.PLEASE NOTE: SPECIAL NECK SIZES AND CONFIGURATIONS WITH OPTIONAL O-RING MATERIALS ARE AVAILABLE. PLEASE CONTACT US FOR DETAILS.