Learn More
Chemglass Life Sciences Reaction Kettle Lid, 300mm, Stainless Steel, 7 Neck, 45/50, Supplied Complete with 45/50 PTFE Inserts and Perfluoro O Rings, 106mm Center to Center Spacing on Necks
SDP
Catalog No. 5000980720
Encompass_Preferred
Reaction vessel lid is constructed of 1/4” thick #316 Stainless Steel with a 1/8” thick PTFE gasket underneath. The only materials exposed to the reaction are PTFE and Perfluoro. Lid has seven openings that accept 45/50 outer PTFE inserts. Inserts are secured to the lid using glass filled PTFE nuts. Lid can be used on any ofCG-1968 or CG-1972 reaction vessels. The flat design reduces the height 200mm than the standard glass reaction vessel lids. PLEASE NOTE: Stainless Steel HEAD DESIGNED FOR USE WITH CG-1968-X AND CG-1968-F SUPPORT FRAMES, AND NOT FOR USE WITH CG-1972-F SUPPORT FRAME. PLEASE NOTE: SPECIAL NECK SIZES AND CONFIGURATIONS WITH OPTIONAL O-RING MATERIALS ARE AVAILABLE.
Catalog No. 50-009-80720 Supplier Chemglass Life Sciences Supplier No. CG1968P100
Reaction vessel lid is constructed of 1/4” thick #316 Stainless Steel with a 1/8” thick PTFE gasket underneath. The only materials exposed to the reaction are PTFE and Perfluoro. Lid has seven openings that accept 45/50 outer PTFE inserts. Inserts are secured to the lid using glass filled PTFE nuts. Lid can be used on any ofCG-1968 or CG-1972 reaction vessels. The flat design reduces the height 200mm than the standard glass reaction vessel lids. PLEASE NOTE: Stainless Steel HEAD DESIGNED FOR USE WITH CG-1968-X AND CG-1968-F SUPPORT FRAMES, AND NOT FOR USE WITH CG-1972-F SUPPORT FRAME. PLEASE NOTE: SPECIAL NECK SIZES AND CONFIGURATIONS WITH OPTIONAL O-RING MATERIALS ARE AVAILABLE.