Learn More
Ace Glass, Inc. Head, 4in Duran O-ring groove flange, 34/45 center, (3)24/40 side necks
SDP
Catalog No. 50979618
Encompass_Preferred
2000mL reaction kettle head with 4 Duran o-ring groove flange, a 34/45 center neck, and (3) 24/40 side necks.
Catalog No. 50-979-618 Supplier Ace Glass, Inc. Supplier No. 802702
2000mL reaction kettle head with 4 Duran o-ring groove flange, a 34/45 center neck, and (3) 24/40 side necks.