Learn More
Sarstedt Inc REACTION TUBE,1.5 ML
Catalog No. NC2215542 Shop All Sarstedt Inc. Products
Encompass_Preferred
Reaction tube, working volume: 1.5 ml, material: PP, transparent, cap: yellow, retaining loop cap, cap attached,500 piece(s)/bag
This item is not returnable. View return policy
Catalog No. NC2215542 Supplier Sarstedt Inc Supplier No. 72.688.004
Reaction tube, working volume: 1.5 ml, material: PP, transparent, cap: yellow, retaining loop cap, cap attached,500 piece(s)/bag