Learn More
Chemglass Life Sciences Chemglass Reaction Vessel, 100 mL, Shallow Dish Bottom (57 mm OD, 90 mm OAH)
SDP
Catalog No. NC2021672
Encompass_Preferred
The Chemglass reaction vessel is a 100 mL glass vessel with a shallow dish bottom designed for small-scale synthesis and automated reactor systems such as Mettler EasyMax. It provides a compact reaction chamber with a 57 mm outer diameter and 90 mm overall height for efficient mixing and heating.
  • 100 mL volume.
  • Outer diameter: 57 mm.
  • Overall height: 90 mm.
  • Shallow dish bottom for improved mixing.
  • Glass construction for chemical compatibility.
  • Compatible with automated reaction platforms.
  • Compact footprint for benchtop and automated workflows.
Catalog No. NC2021672 Supplier Chemglass Life Sciences Supplier No. CGMR100V1
The Chemglass reaction vessel is a 100 mL glass vessel with a shallow dish bottom designed for small-scale synthesis and automated reactor systems such as Mettler EasyMax. It provides a compact reaction chamber with a 57 mm outer diameter and 90 mm overall height for efficient mixing and heating.
  • 100 mL volume.
  • Outer diameter: 57 mm.
  • Overall height: 90 mm.
  • Shallow dish bottom for improved mixing.
  • Glass construction for chemical compatibility.
  • Compatible with automated reaction platforms.
  • Compact footprint for benchtop and automated workflows.