Learn More
Chemglass Life Sciences Reaction Vessel, 50mL, One-Piece, 5-Neck, Equivalent to Mettler Easymax Part # 51161792
SDP
Catalog No. 501656788
Encompass_Preferred
One-Piece Reaction Vessel, 5-Nk, 50mL: 19/22 Outer CN, (1) 10/18 Outer SN, (1) 14/20 Outer SN, and (2) 19/22 Outer SN's. Vessel is graduated from 10 to 50mL. Body outside diameter is 43mm. Equivalent to Mettler EasyMax 102/402 Part # 5116172.Mettler Easymax® is a registered trademark of Mettler Toledo.
Catalog No. 50-165-6788 Supplier Chemglass Life Sciences Supplier No. CGMR50V1P
One-Piece Reaction Vessel, 5-Nk, 50mL: 19/22 Outer CN, (1) 10/18 Outer SN, (1) 14/20 Outer SN, and (2) 19/22 Outer SN's. Vessel is graduated from 10 to 50mL. Body outside diameter is 43mm. Equivalent to Mettler EasyMax 102/402 Part # 5116172.Mettler Easymax® is a registered trademark of Mettler Toledo.