Learn More
Globe Scientific Reaction Vessels for Abbot Architect Series Analyzers
Shop All Globe Scientific ProductsDescription
- For use with Abbot Architect Series Analyzers
- Designed specifically to work with these products
- Polypropylene construction, natural color
Specifications
Specifications
| Type | Reaction Vessel |
| Color | Natural |
| Quantity | 2000 |
| Material | Polypropylene |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.