Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Chemglass Reaction Vessel Lid, 6-Neck, 300 mm Flange, 45/50 and 80 mm Ports (angled 20°) - fits 300 mm flanged reaction vessels
SDP

Supplier:  Chemglass Life Sciences CG1968A20

Encompass_Preferred

Chemglass 6-neck reaction vessel lid designed for 300 mm flanged reaction vessels. The lid provides multiple standardized necks and flat o-ring flanges to accept condensers, thermometers, addition funnels, and other standard laboratory fittings, and includes angled flanges for powder addition.

  • Six neck configuration with standardized joints and o-ring flanges.
  • Neck sizes: one center 45/50 and three vertical 45/50 side necks.
  • Two 80 mm flat o-ring flanges angled 20° for powder or solids addition.
  • Flange diameter: 300 mm suitable for 300 mm flanged reaction vessels.
  • Material: not specified on manufacturer page.

Catalog No. 50-273-3556


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.