Learn More
Chemglass Life Sciences Reaction Vessel Lid Complete, SS, 400mm, 7-Neck, Large Addition Port, Perfluoro O-Rings
SDP
Catalog No. 5000980721
Encompass_Preferred
Reaction vessel lid is constructed of 1/4” thick #316 Stainless Steel with a 1/8” thick PTFE gasket underneath. The only material exposed to the reaction is PTFE. Lid has seven openings that accept 45/50 outer PTFE inserts. One neck is a bulk addition port is 80mm reaction flange with o-ring groove. Inserts are secured to the lid using glass filed PTFE nuts. Lid can be used on any ofCG-1968-W style reactors. The flat design reduces the height over the standard glass reaction vessel lids. PLEASE NOTE: SPECIAL NECK SIZES AND CONFIGURATIONS ARE AVAILABLE,.
Catalog No. 50-009-80721 Supplier Chemglass Life Sciences Supplier No. CG1968P400
Reaction vessel lid is constructed of 1/4” thick #316 Stainless Steel with a 1/8” thick PTFE gasket underneath. The only material exposed to the reaction is PTFE. Lid has seven openings that accept 45/50 outer PTFE inserts. One neck is a bulk addition port is 80mm reaction flange with o-ring groove. Inserts are secured to the lid using glass filed PTFE nuts. Lid can be used on any ofCG-1968-W style reactors. The flat design reduces the height over the standard glass reaction vessel lids. PLEASE NOTE: SPECIAL NECK SIZES AND CONFIGURATIONS ARE AVAILABLE,.