Learn More
Chemglass Life Sciences 500mL Reaction Vessel, Jacketed, Graduated Both Sides, 100mm Flange, Flat Flange Inlet/Outlet, CG-911-A Drain Valve
SDP
Catalog No. 50143608
Encompass_Preferred
500mL Reaction Vessel, Jacketed, Graduated Both Sides, 100mm Flange, Flat Flange Inlet/Outlet, CG-911-A Drain Valve.Features:• Tool-Free™ Jacket Connection Design Allows Hose to be Attached or Removed by Hand Without Using Tools• Chemically Resistant Perfluoro O-Rings on All CG-911-A and CG-511 Drain Valves• Sidearm on Lower Drain Valve can be Used with a Standard 1/2” or 3/4” TFE, PFA or Polypropylene Compression Fitting• All Vessels are Serial Numbered
Catalog No. 50-143-608 Supplier Chemglass Life Sciences Supplier No. CG1929X13
500mL Reaction Vessel, Jacketed, Graduated Both Sides, 100mm Flange, Flat Flange Inlet/Outlet, CG-911-A Drain Valve.Features:• Tool-Free™ Jacket Connection Design Allows Hose to be Attached or Removed by Hand Without Using Tools• Chemically Resistant Perfluoro O-Rings on All CG-911-A and CG-511 Drain Valves• Sidearm on Lower Drain Valve can be Used with a Standard 1/2” or 3/4” TFE, PFA or Polypropylene Compression Fitting• All Vessels are Serial Numbered