Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach™ Reaction Vessels with Screw Caps

Catalog No. 501997887 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Quantity:
5 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-7887 5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-7887 Supplier Hach™ Supplier No. LZP065
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Hach™ Reaction Vessels with Screw Caps are 20mm diameter glass vials for sample reactions/digestions.

  • Designed for use with TNTplus Vial Test systems
  • Pack of 5 tubes

Order Info

dimProductType

TRUSTED_SUSTAINABILITY

Specifications

Analytes Boron
Color Clear, red cap
Description Glass vials
For Use With (Application) For sample reactions and digestions
For Use With (Equipment) TNTplus Vial Test systems
Includes Screw caps
Quantity 5 Each
Size 20 mm Dia
Type Reaction Vessel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.