Learn More
Chemglass Life Sciences Storage Vial, 20mL, Complete, Clear, 28 x 57mm, 24-400 White Solid Cap, with Bonded Silicone/PTFE Liner
SDP
Catalog No. 501215210
Encompass_Preferred
Storage Vial, 20mL, Complete, Clear, 28 x 57mm, 24-400 White Solid Cap, TFE Septa. For storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Caps are supplied installed on the vials and all sizes have the TFE lined silicone SURE-LINK septa.
Catalog No. 50-121-5210 Supplier Chemglass Life Sciences Supplier No. CG490803
Storage Vial, 20mL, Complete, Clear, 28 x 57mm, 24-400 White Solid Cap, TFE Septa. For storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identification. Caps are supplied installed on the vials and all sizes have the TFE lined silicone SURE-LINK septa.