Learn More
Chemglass Life Sciences Storage Vial, 8mL, Complete, Clear, 17 x 60mm, 15-425 Green Solid PTFE Lined Caps
SDP
Catalog No. NC0054656
Encompass_Preferred
Storage/Reaction Vial, 8mL, Complete, Clear, 17 x 60mm, 15-425 Green Solid Cap, TFE Septa, Case of 100
Catalog No. NC0054656 Supplier Chemglass Life Sciences Supplier No. CG490805
Storage/Reaction Vial, 8mL, Complete, Clear, 17 x 60mm, 15-425 Green Solid Cap, TFE Septa, Case of 100