Learn More
Chemglass Life Sciences Vial, 8mL (2 Dram), Complete with Green Solid Top Cap, TFE Septa, 17x60mm, 15-425 Thread
SDP
Catalog No. 501531776
Encompass_Preferred
Vial, 8mL (2 Dram), Complete with Green Solid Top Cap, TFE Septa, 17x60mm, 15-425 Thread. Packaged in a clear PETG tray complete with a slide or lift off dust cover. Vials are separated by a SBS partition to eliminate glass to glass contact. Caps are supplied complete in the package, not attached. Entire package is recyclable. Vials are made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585. The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for vials along with most all laboratory glassware. Polypropylene caps have a septa bonded to the cap, using the Chemglass SURE-Link Process. The inert process bonds the PTFE Faced Silicone septa directly to the cap without adhesives, making the caps autoclavable and able to withstand multiple injections.
Catalog No. 50-153-1776 Supplier Chemglass Life Sciences Supplier No. CG490409
Vial, 8mL (2 Dram), Complete with Green Solid Top Cap, TFE Septa, 17x60mm, 15-425 Thread. Packaged in a clear PETG tray complete with a slide or lift off dust cover. Vials are separated by a SBS partition to eliminate glass to glass contact. Caps are supplied complete in the package, not attached. Entire package is recyclable. Vials are made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585. The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for vials along with most all laboratory glassware. Polypropylene caps have a septa bonded to the cap, using the Chemglass SURE-Link Process. The inert process bonds the PTFE Faced Silicone septa directly to the cap without adhesives, making the caps autoclavable and able to withstand multiple injections.