Learn More
Medchemexpress LLC Reactive Blue 19 | 2580-78-1 | 100% | 10 G
SDP
Catalog No. 5000410863
Encompass_Preferred
Reactive Blue 19 is an anthraquinone dye used in the textile industry. It can also serve as a starting material for producing polymer dyes. It is described as a toxic and undegradable organic pollutant that poses a threat to human health and aquatic life.
  • Anthraquinone dye
  • Used in the textile industry
  • Can be a starting material for polymer dyes
  • Toxic and undegradable organic pollutant
  • For research use only
Catalog No. 50-004-10863 Supplier Medchemexpress LLC Supplier No. HYD033110G
Add to Cart
Add to Cart
Reactive Blue 19 is an anthraquinone dye used in the textile industry. It can also serve as a starting material for producing polymer dyes. It is described as a toxic and undegradable organic pollutant that poses a threat to human health and aquatic life.
  • Anthraquinone dye
  • Used in the textile industry
  • Can be a starting material for polymer dyes
  • Toxic and undegradable organic pollutant
  • For research use only