Learn More
Bioworld REACTIVE BLUE-4 SEPAROPORE
Supplier: Bioworld 201810971
This item has a minimum order quantity of 2 per supplier requirements. REACTIVE BLUE-4 SEPAROPORE (AGAROSE) 4B-CL: Reactive Blue 4 bound to Cross-linked 4% agarose beads (Separopore 4B-CL). Specifications Matrix: Cross-linked agarose, 4% Dye:Reactive Blue 4 Ligand coupling method: Triazine coupling ...