Learn More
Bioworld REACTIVE BLUE-4 SEPAROPORE
Catalog No. 50197912
Encompass
REACTIVE BLUE-4 SEPAROPORE (AGAROSE) 4B-CL: Reactive Blue 4 bound to Cross-linked 4% agarose beads (Separopore 4B-CL). Specifications Matrix: Cross-linked agarose, 4% Dye:Reactive Blue 4 Ligand coupling method: Triazine coupling ...
Catalog No. 50-197-912 Supplier Bioworld Supplier No. 201810972
REACTIVE BLUE-4 SEPAROPORE (AGAROSE) 4B-CL: Reactive Blue 4 bound to Cross-linked 4% agarose beads (Separopore 4B-CL). Specifications Matrix: Cross-linked agarose, 4% Dye:Reactive Blue 4 Ligand coupling method: Triazine coupling ...