Learn More
Biotium Reactive DNA RNA Binding Dyes PCR and DNA Amplification DNA Binding Dyes Chemical Reagents 1/EA
Catalog No. 5000501561
Encompass_Preferred
Methylene Blue (MB) is a commonly used redox indicator in nucleic acid research MB-Maleimide can be used to label molecules containing free thiol groups
Catalog No. 50-005-01561 Supplier Biotium Supplier No. 40118
Methylene Blue (MB) is a commonly used redox indicator in nucleic acid research MB-Maleimide can be used to label molecules containing free thiol groups