Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reactive Yellow 86, Practical grade, MP Biomedicals™

Catalog No. ICN19547925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19547925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19547925 Supplier MP Biomedicals, Inc Supplier No. 0219547925
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Practical grade Reactive Yellow 86 dye suitable for laboratory use.

  • Grade: Practical
  • Physical Form: Powder
  • Molecular Formula: C18H14Cl2N8Na2O9S2
  • Molecular Weight: 667.357 g/mol
  • CAS Number: 61951-86-8
  • Recommended Storage: Store at room temperature
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61951-86-8
Molecular Formula C18H14Cl2N8Na2O9S2
Molecular Weight (g/mol) 667.357
InChI Key JUNRGNKPIKIXLL-UHFFFAOYSA-L
PubChem CID 57369787
IUPAC Name disodium;4-[2-(5-carbamoyl-1-ethyl-4-methyl-2,6-dioxopyridin-3-ylidene)hydrazinyl]-6-[(4,6-dichloro-1,3,5-triazin-2-yl)amino]benzene-1,3-disulfonate
SMILES CCN1C(=O)C(=C(C(=NNC2=C(C=C(C(=C2)NC3=NC(=NC(=N3)Cl)Cl)S(=O)(=O)[O-])S(=O)(=O)[O-])C1=O)C)C(=O)N.[Na+].[Na+]

Specifications

Color Yellow
Quantity 25 g
Grade Practical
Chemical Name or Material Reactive Yellow 86
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.