Learn More
Chemglass Life Sciences Chemglass Reaction Vessel Lid, 6-Neck, 250 mL (CG-MR-250L2)
SDP
Catalog No. NC2174923
Encompass_Preferred
Chemglass CG-MR-250L2 is a six-neck reaction vessel lid designed for 250 mL laboratory reaction vessels and compatible with Multimax-style reaction systems. It provides multiple standard joints for attaching condensers, thermometers, and probes, and mounts on a 98 mm flange for secure sealing.
  • Six-neck configuration for multiple attachments.
  • Designed for 250 mL reaction vessels with a 98 mm flange.
  • Center neck and side necks accept standard tapered joints for common lab glassware and probes.
  • Includes multiple standard joint sizes to support condensers and instrumentation.
  • Manufactured laboratory glassware for chemical compatibility and durability.
This item is not returnable. View return policy
Catalog No. NC2174923 Supplier Chemglass Life Sciences Supplier No. CGMR250L2
Chemglass CG-MR-250L2 is a six-neck reaction vessel lid designed for 250 mL laboratory reaction vessels and compatible with Multimax-style reaction systems. It provides multiple standard joints for attaching condensers, thermometers, and probes, and mounts on a 98 mm flange for secure sealing.
  • Six-neck configuration for multiple attachments.
  • Designed for 250 mL reaction vessels with a 98 mm flange.
  • Center neck and side necks accept standard tapered joints for common lab glassware and probes.
  • Includes multiple standard joint sizes to support condensers and instrumentation.
  • Manufactured laboratory glassware for chemical compatibility and durability.