Learn More
Ace Glass, Inc. Reactor, 20 liter flush valve jacketed vessel, 200mmflask head (45/50 center, (3)45/50, (1)60mm port)an IKA Eurostar 60 Control overhead stirrer, shaft and agitators
SDP
Catalog No. 50977149
Encompass
Reactor, 20 liter flush valve jacketed vessel, 200mmflask head (45/50 center, (3)45/50, (1)60mm port)an IKA Eurostar 60 Control overhead stirrer, shaft and agitators
Catalog No. 50-977-149 Supplier Ace Glass, Inc. Supplier No. 10001201
Add to Cart
Add to Cart
Request a Quote
Reactor, 20 liter flush valve jacketed vessel, 200mmflask head (45/50 center, (3)45/50, (1)60mm port)an IKA Eurostar 60 Control overhead stirrer, shaft and agitators