Learn More
Hach Company Reducing adapter, 16 mm to 13 mm vials, 5 pieces
Supplier:  Hach Company 2895805
Encompass
These aluminum tubes are inserted into the 16 mm vial wells of existing reactors, to adapt them to the 13 mm TNTplus vials. The adapters can be removed (once the heater block has cooled) if the customer needs to use 16 mm vial chemistries.
Catalog No. 50-199-6772