Learn More
Ace Glass, Inc. 500mL pressure vessel body, jacketed, 100mm flange, 0-8mmvalve, #11 Ace-Thred inlets/outlets for use with 3/8in .
SDP
Catalog No. 50972917
Encompass_Preferred
500mL pressure vessel body, jacketed, 100mm flange, 0-8mmvalve, #11 Ace-Thred inlets/outlets for use with 3/8in .
Catalog No. 50-972-917 Supplier Ace Glass, Inc. Supplier No. 6429314
500mL pressure vessel body, jacketed, 100mm flange, 0-8mmvalve, #11 Ace-Thred inlets/outlets for use with 3/8in .