Learn More
Ace Glass, Inc. 3000mL pressure vessel body, jacketed, 150mm flange, round bottom, #15 Ace-Thred inlet/outlet, use with 1/2in ID tubing
SDP
Catalog No. 50972897
Encompass_Preferred
3000mL pressure vessel body, jacketed, 150mm flange, round bottom, #15 Ace-Thred inlet/outlet, use with 1/2in ID tubing
Catalog No. 50-972-897 Supplier Ace Glass, Inc. Supplier No. 6427323
Add to Cart
Add to Cart
Request a Quote
3000mL pressure vessel body, jacketed, 150mm flange, round bottom, #15 Ace-Thred inlet/outlet, use with 1/2in ID tubing