Learn More
Ace Glass, Inc. 3000mL pressure vessel body, jacketed, 150mm flange, 0-10mm valve, 28/15 ball joint bottom outlet, 28/15 ball joints on jacket inlet/outlet.
SDP
Catalog No. 50972911
Encompass_Preferred
3000mL pressure vessel body, jacketed, 150mm flange, 0-10mm valve, 28/15 ball joint bottom outlet, 28/15 ball joints on jacket inlet/outlet.
Catalog No. 50-972-911 Supplier Ace Glass, Inc. Supplier No. 642928
Add to Cart
Add to Cart
Request a Quote
3000mL pressure vessel body, jacketed, 150mm flange, 0-10mm valve, 28/15 ball joint bottom outlet, 28/15 ball joints on jacket inlet/outlet.