Learn More
Ace Glass, Inc. 1000mL pressure vessel body, one-piece, jacketed, #25 center neck, (3) #15 and (1) #7 side necks, 0-8mm 28/15 ball joint bottom outlet, 28/15 O-ring ball joint on jacket inlet/outlet
SDP
Catalog No. 50978725
Encompass_Preferred
1000mL pressure vessel body, one-piece, jacketed, #25 center neck, (3) #15 and (1) #7 side necks, 0-8mm 28/15 ball joint bottom outlet, 28/15 O-ring ball joint on jacket inlet/outlet
Catalog No. 50-978-725 Supplier Ace Glass, Inc. Supplier No. 643921
1000mL pressure vessel body, one-piece, jacketed, #25 center neck, (3) #15 and (1) #7 side necks, 0-8mm 28/15 ball joint bottom outlet, 28/15 O-ring ball joint on jacket inlet/outlet