Learn More
Ace Glass, Inc. 2000mL pressure vessel body, one-piece, jacketed, #25 center neck, (3) #15 and (1) #7 side necks, 0-8mm 28/15 ball joint bottom outlet, 28/15 O-ring ball joint on jacket inlet/outlet
SDP
Catalog No. 50972957
Encompass_Preferred
2000mL pressure vessel body, one-piece, jacketed, #25 center neck, (3) #15 and (1) #7 side necks, 0-8mm 28/15 ball joint bottom outlet, 28/15 O-ring ball joint on jacket inlet/outlet
Catalog No. 50-972-957 Supplier Ace Glass, Inc. Supplier No. 643925
2000mL pressure vessel body, one-piece, jacketed, #25 center neck, (3) #15 and (1) #7 side necks, 0-8mm 28/15 ball joint bottom outlet, 28/15 O-ring ball joint on jacket inlet/outlet