Learn More
Ace Glass, Inc. 2000mL pressure vessel body, one-piece, jacketed, #25 center neck, (3) #15 and (1) #7 side necks, 0-8mm 28/15 ball joint bottom outlet, #11 Ace-Thred connection on jacket inlet/outlet for use with 3/8in ID tubing.
SDP
Catalog No. 50972965
Encompass_Preferred
2000mL pressure vessel body, one-piece, jacketed, #25 center neck, (3) #15 and (1) #7 side necks, 0-8mm 28/15 ball joint bottom outlet, #11 Ace-Thred connection on jacket inlet/outlet for use with 3/8in ID tubing.
Catalog No. 50-972-965 Supplier Ace Glass, Inc. Supplier No. 6439325
Add to Cart
Add to Cart
2000mL pressure vessel body, one-piece, jacketed, #25 center neck, (3) #15 and (1) #7 side necks, 0-8mm 28/15 ball joint bottom outlet, #11 Ace-Thred connection on jacket inlet/outlet for use with 3/8in ID tubing.