Learn More
Ace Glass, Inc. 3000mL pressure vessel body, one-piece, jacketed, #25 center neck, (4) #15 and (1) #7 side necks, 0-10mm 28/15 ball joint bottom outlet, 28/15 O-ring ball joint on jacket inlet/outlet
SDP
Catalog No. 50972958
Encompass_Preferred
3000mL pressure vessel body, one-piece, jacketed, #25 center neck, (4) #15 and (1) #7 side necks, 0-10mm 28/15 ball joint bottom outlet, 28/15 O-ring ball joint on jacket inlet/outlet
Catalog No. 50-972-958 Supplier Ace Glass, Inc. Supplier No. 643929
3000mL pressure vessel body, one-piece, jacketed, #25 center neck, (4) #15 and (1) #7 side necks, 0-10mm 28/15 ball joint bottom outlet, 28/15 O-ring ball joint on jacket inlet/outlet