Learn More
Ace Glass, Inc. 3000mL pressure vessel body, one-piece, jacketed, #25 center, (4) #15, (1) #7 side necks, round bottom, #15 Ace-Thred connections on jacket inlet/outlet, use 1/2in tubing.
SDP
Catalog No. 50978723
Encompass_Preferred
3000mL one-piece jacketed round bottom pressure vessel with a #25 center neck, (4) #15 and (1) #7 side necks, and a #15 Ace-Thred connection for 3/8 tubing on the jacket inlet/outlet. Vessel is part of pressure reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Rated at 45psi at 100 degrees Centigrade.
Catalog No. 50-978-723 Supplier Ace Glass, Inc. Supplier No. 6438324
Add to Cart
Add to Cart
3000mL one-piece jacketed round bottom pressure vessel with a #25 center neck, (4) #15 and (1) #7 side necks, and a #15 Ace-Thred connection for 3/8 tubing on the jacket inlet/outlet. Vessel is part of pressure reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Rated at 45psi at 100 degrees Centigrade.