Learn More
Ace Glass, Inc. 5000mL pressure vessel body, one-piece, jacketed, #25 center neck, (4) #15 and (1) #7 side necks, 0-10mm 28/15 ball joint bottom outlet, #15 Ace-Thred connection on jacket inlet/outlet for use with 1/2in ID tubing.
SDP
Catalog No. 50972966
Encompass_Preferred
5000mL pressure vessel body, one-piece, jacketed, #25 center neck, (4) #15 and (1) #7 side necks, 0-10mm 28/15 ball joint bottom outlet, #15 Ace-Thred connection on jacket inlet/outlet for use with 1/2in ID tubing.
Catalog No. 50-972-966 Supplier Ace Glass, Inc. Supplier No. 6439334
Add to Cart
Add to Cart
Request a Quote
5000mL pressure vessel body, one-piece, jacketed, #25 center neck, (4) #15 and (1) #7 side necks, 0-10mm 28/15 ball joint bottom outlet, #15 Ace-Thred connection on jacket inlet/outlet for use with 1/2in ID tubing.