Learn More
Ace Glass, Inc. 5000mL pressure vessel body, one-piece, jacketed, #25 center, (4) #15, (1) #7 side necks, round bottom, 28/15 O-ring ball joints on inlet/outlet
SDP
Catalog No. 50972944
Encompass_Preferred
5000mL one-piece jacketed round bottom pressure vessel with a #25 center neck, (4) #15 and (1) #7 side necks, and a 28/15 o-ring ball joint on the inlet/outlet. Vessel is part of pressure reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Rated at 45psi at 100 degrees Centigrade.
Catalog No. 50-972-944 Supplier Ace Glass, Inc. Supplier No. 643829
5000mL one-piece jacketed round bottom pressure vessel with a #25 center neck, (4) #15 and (1) #7 side necks, and a 28/15 o-ring ball joint on the inlet/outlet. Vessel is part of pressure reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Rated at 45psi at 100 degrees Centigrade.