Learn More
Ace Glass, Inc. Reactor body, glass only, photochem, 1000mL
SDP
Catalog No. 50975800
Encompass_Preferred
1000mL glass, unjacketed photochem reactor vessel designed for use with ACE quartz and glass immersion wells. Vessel includes a flat bottom that will accept the use of a stir magnet.
Catalog No. 50-975-800 Supplier Ace Glass, Inc. Supplier No. 786320
1000mL glass, unjacketed photochem reactor vessel designed for use with ACE quartz and glass immersion wells. Vessel includes a flat bottom that will accept the use of a stir magnet.