Learn More
Ace Glass, Inc. Reactor body, glass only, photochem, 250mL
SDP
Catalog No. 50975798
Encompass_Preferred
250mL glass, unjacketed photochem reactor vessel designed for use with ACE quartz and glass immersion wells. Vessel includes a flat bottom that will accept the use of a stir magnet.
Catalog No. 50-975-798 Supplier Ace Glass, Inc. Supplier No. 786316
250mL glass, unjacketed photochem reactor vessel designed for use with ACE quartz and glass immersion wells. Vessel includes a flat bottom that will accept the use of a stir magnet.