Learn More
Ace Glass, Inc. 1000mL pressure vessel body, one-piece, #25 center, (3) #15, (1 )#7 side necks, round bottom
SDP
Catalog No. 50978715
Encompass_Preferred
1000mL round bottom pressure vessel body. Vessel has a #25 Ace-Thred center neck, 3 #15 Ace-Thred side necks, and 1 #7 Ace-Thred side neck. Rated 45 PSI at 100C.
Catalog No. 50-978-715 Supplier Ace Glass, Inc. Supplier No. 643609
1000mL round bottom pressure vessel body. Vessel has a #25 Ace-Thred center neck, 3 #15 Ace-Thred side necks, and 1 #7 Ace-Thred side neck. Rated 45 PSI at 100C.