Learn More
Ace Glass, Inc. 500mL one-piece pressure vessel body, #25 center neck, (3)#15 and (1) #7 side necks, 0-8mm 28/15 ball joint bottom outlet
SDP
Catalog No. 50972933
Encompass_Preferred
500mL one-piece pressure vessel body featuring a #25 Ace-Thred center neck, three #15 and one #7 side necks. Vessel also features a 0-8mm, 28/15 ball joint bottom outlet.
Catalog No. 50-972-933 Supplier Ace Glass, Inc. Supplier No. 643707
500mL one-piece pressure vessel body featuring a #25 Ace-Thred center neck, three #15 and one #7 side necks. Vessel also features a 0-8mm, 28/15 ball joint bottom outlet.