Learn More
Ace Glass, Inc. 500mL pressure vessel body, one-piece, #25 center, (3) #15, (1)#7 side necks, round bottom
SDP
Catalog No. 50972926
Encompass_Preferred
500mL round bottom pressure vessel body. Vessel has a #25 Ace-Thred center neck, 3 #15 Ace-Thred side necks, and 1 #7 Ace-Thred side neck. Rated 45 PSI at 100C.
Catalog No. 50-972-926 Supplier Ace Glass, Inc. Supplier No. 643606
500mL round bottom pressure vessel body. Vessel has a #25 Ace-Thred center neck, 3 #15 Ace-Thred side necks, and 1 #7 Ace-Thred side neck. Rated 45 PSI at 100C.