Learn More
Ace Glass, Inc. 5000mL one-piece pressure vessel body, #25 center neck, (4)#15 and (1) #7 side necks, 0-10mm 28/15 ball joint bottom outlet
SDP
Catalog No. 50972936
Encompass_Preferred
5000mL one-piece pressure vessel body featuring a #25 Ace-Thred center neck, four #15 and one #7 side necks. Vessel also features a 0-10mm, 28/15 ball joint bottom outlet.
Catalog No. 50-972-936 Supplier Ace Glass, Inc. Supplier No. 643724
5000mL one-piece pressure vessel body featuring a #25 Ace-Thred center neck, four #15 and one #7 side necks. Vessel also features a 0-10mm, 28/15 ball joint bottom outlet.