Learn More
METTLER TOLEDO REACTOR HP60-V SS 300C RC1
Catalog No. 502741649 Shop All METTLER TOLEDO Products
Encompass
Reactor HP60-V, SS, 300°C (RC1mx)
Catalog No. 50-274-1649 Supplier METTLER TOLEDO Supplier No. 30425630
Add to Cart
Add to Cart
Request a Quote
Reactor HP60-V, SS, 300°C (RC1mx)