Learn More
Ace Glass, Inc. 2000mL, two-piece pressure reactor system, complete with glass body, 100mm head with (1) #7 and (5) #15 necks, addition funnel, condenser, manifold with gauge and rupture disc, stirring and temperature control assemblies
SDP
Catalog No. 50978696
Encompass
2000mL, two-piece pressure reactor system, complete with glass body, 100mm head with (1) #7 and (5) #15 necks, addition funnel, condenser, manifold with gauge and rupture disc, stirring and temperature control assemblies
Catalog No. 50-978-696 Supplier Ace Glass, Inc. Supplier No. 6423215
Add to Cart
Add to Cart
Request a Quote
2000mL, two-piece pressure reactor system, complete with glass body, 100mm head with (1) #7 and (5) #15 necks, addition funnel, condenser, manifold with gauge and rupture disc, stirring and temperature control assemblies
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our missing translation for 'privacyPolicy'.