Learn More
Ace Glass, Inc. 3000mL, two-piece pressure reactor system, complete with glass body, 150mm head with (1) #7 and (5) #15 necks, addition funnel, condenser, manifold with gauge and rupture disc, stirring and temperature control assemblies