Learn More
Ace Glass, Inc. 2000mL, one-piece pressure reactor with #25 center, (3)#15, (1)#7 side necks, 0-8mm valve and 28/15 ball joint bottom outlet, complete with addition funnel, condenser, pressure manifold, stirring and temperature control assemblies
SDP
Catalog No. 50978719
Encompass
2000mL one-piece pressure reactor featuring a #25 Ace-Thred center neck, three #15 and one #7 side necks. Vessel also features a 0-8mm, 28/15 ball joint bottom outlet. Reactor is complete with an addition funnel, condenser, heating mantle, pressure manifold, stir shaft and temperature controller. Unit will perform positive pressure reactions up to 45psi at 100 C.
Catalog No. 50-978-719 Supplier Ace Glass, Inc. Supplier No. 6437241
Add to Cart
Add to Cart
Request a Quote
2000mL one-piece pressure reactor featuring a #25 Ace-Thred center neck, three #15 and one #7 side necks. Vessel also features a 0-8mm, 28/15 ball joint bottom outlet. Reactor is complete with an addition funnel, condenser, heating mantle, pressure manifold, stir shaft and temperature controller. Unit will perform positive pressure reactions up to 45psi at 100 C.