Learn More
Ace Glass, Inc. 2000mL, one-piece pressure reactor with #25 center, (3)#15, (1)#7 side necks, complete with addition funnel, condenser, pressure manifold, stirring and temperature control assemblies
SDP
Catalog No. 50972931
Encompass
2000mL one-piece pressure reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Flask features #25 Ace-Thred center neck, 3 #15 Ace-Thred side necks, and 1 #7 Ace-Thred side neck. System is complete with addition funnel, condenser, pressure manifold, stirring and temperature control assemblies. Rated 45 PSI at 100C.
Catalog No. 50-972-931 Supplier Ace Glass, Inc. Supplier No. 6436239
Add to Cart
Add to Cart
Request a Quote
2000mL one-piece pressure reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Flask features #25 Ace-Thred center neck, 3 #15 Ace-Thred side necks, and 1 #7 Ace-Thred side neck. System is complete with addition funnel, condenser, pressure manifold, stirring and temperature control assemblies. Rated 45 PSI at 100C.