Learn More
Ace Glass, Inc. 5000mL, one-piece jacketed pressure reactor with #25 center, (4)#15, (1)#7 side necks, 0-10mm valve and 28/15 ball joint bottom outlet, 28/15 ball joint inlet/outlet, complete with addition funnel, condenser, manifold & stirring assemblies
SDP
Supplier:  Ace Glass, Inc. 6439257
Encompass
Complete, one-piece 5000mL jacketed pressure reactor is designed to perform low to moderate positive pressure reactions, synthesis, and catalysis. Flask has a #25 center neck, (4) #15 side necks, and (1) #7 side neck. Flask bottom outlet is 0-10mm 28/15 ball joint. Jacket inlet/outlet has a 28/15 o-ring ball joint. Rated 45 PSI at 100 degrees C. Reactor system includes addition funnel, condenser, manifold, and stirring assemblies.
Catalog No. 50-978-727
Add to Cart
Request a Quote