Learn More
Ace Glass, Inc. 5000mL, one-piece pressure reactor with #25 center, (4)#15, (1)#7 side necks, 0-10mm valve and 28/15 ball joint bottom outlet, complete with addition funnel, condenser, pressure manifold, stirring and temperature control assemblies
SDP
Supplier:  Ace Glass, Inc. 6437246
Encompass
5000mL one-piece pressure reactor featuring a #25 Ace-Thred center neck, four #15 and one #7 side necks. Vessel also features a 0-10mm, 28/15 ball joint bottom outlet. Reactor is complete with an addition funnel, condenser, heating mantle, pressure manifold, stir shaft and temperature controller. Unit will perform positive pressure reactions up to 45psi at 100 C.
Catalog No. 50-972-940
Add to Cart
Request a Quote